Product Details

SNP ID
rs4230240
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.14:30039970 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
TCTTACAAAGAGAAATTTAATATTA[A/G]ATGAGAGGTTGAACCAGGCTTAAAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
Cacna1d PubMed Links

Gene Details

Gene
Cacna1d
Gene Name
calcium channel, voltage-dependent, L type, alpha 1D subunit
There are no transcripts associated with this gene.

Gene
Chdh
Gene Name
choline dehydrogenase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136240.1 5156 UTR 3 NP_001129712.1
NM_172264.2 5156 UTR 3 NP_758468.2
NM_175343.5 5156 UTR 3 NP_780552.1

View Full Product Details