Product Details

SNP ID
rs13476103
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.1:127959819 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
GAGCTCCTGTGCGTCCACACCACAG[C/T]GCTGACACACACCATGTTCTGTTGC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
Gm28800 PubMed Links

Gene Details

Gene
Gm28800
Gene Name
predicted gene 28800
There are no transcripts associated with this gene.

Gene
Zranb3
Gene Name
zinc finger, RAN-binding domain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001285945.1 2957 Missense Mutation NP_001272874.1
NM_027678.3 2957 Missense Mutation NP_081954.1

View Full Product Details