Product Details

SNP ID
rs13481653
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.12:111710311 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
CCTTGTGTGGGGGGTGCTCCTCACA[C/T]GGGAACAGCTTCCTCTTCTCGAGGG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
Apopt1 PubMed Links

Gene Details

Gene
Apopt1
Gene Name
apoptogenic, mitochondrial 1
There are no transcripts associated with this gene.

Gene
Bag5
Gene Name
BCL2-associated athanogene 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_027404.2 1315 Silent Mutation NP_081680.1

View Full Product Details