Product Details

SNP ID
rs13481474
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.12:57542267 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
GGAGAAGGGGTGATTAAAGGAGTAG[C/T]GGGGATCCCCTTTCAGATGCAGCTG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
Foxa1 PubMed Links

Gene Details

Gene
Foxa1
Gene Name
forkhead box A1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_008259.3 1552 Missense Mutation NP_032285.2

View Full Product Details