Product Details
- SNP ID
-
rs29344239
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:8332081 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TAACTTTGATGTAAATCTTGAAATT[A/G]GGAAAGTTCTTTCAGCTTTGTTCCT
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Ust
PubMed Links
Gene Details
- Gene
- Ust
- Gene Name
- uronyl-2-sulfotransferase
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_177387.3 |
|
Intron |
|
|
NP_796361.2 |
View Full Product Details