Product Details

SNP ID
rs3664667
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.14:66745348 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
CACCTTATCAAAGCAGAGCGAGTAG[G/A]ATCTCCTGATTGGTTGCCCTCCATG
Phenotype
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
Adra1a PubMed Links

Gene Details

Gene
Adra1a
Gene Name
adrenergic receptor, alpha 1a
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271759.1 Intron NP_001258688.1
NM_001271760.1 Intron NP_001258689.1
NM_001271761.1 Intron NP_001258690.1
NM_013461.4 Intron NP_038489.2

View Full Product Details