Product Details
- SNP ID
-
rs13482507
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:31441369 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTCCGAAAGTCCTATCATGCCATTC[C/T]TTCTAGATTCTCTGGATGGGAAACA
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Ropn1l
PubMed Links
Gene Details
- Gene
- Ropn1l
- Gene Name
- ropporin 1-like
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_145852.2 |
751 |
Missense Mutation |
|
|
NP_665851.2 |
View Full Product Details