Product Details
- SNP ID
-
rs13479650
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:23770632 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACAATTCATAACCTGCAGTCTCTGC[C/T]TGTTGGGGTTTATAGCCATGTCTTC
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Zmat4
PubMed Links
Gene Details
- Gene
- Zmat4
- Gene Name
- zinc finger, matrin type 4
View Full Product Details