Product Details
- SNP ID
-
rs13476439
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:38111118 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TATTCTACTCCAACAACCAAACTTT[C/T]ACACACTGATAAGAAGAAGTATCAA
- Phenotype
-
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
Dennd1a
PubMed Links
Gene Details
- Gene
- Dennd1a
- Gene Name
- DENN/MADD domain containing 1A
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_146122.3 |
|
Intron |
|
|
NP_666234.3 |
View Full Product Details