Product Details
- SNP ID
-
rs3698283
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:43184798 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TAGTGAGAGCCTCTGAGACTTGATA[C/T]GTGTTCCTCTTCAGATCTGTACCTA
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Unc13b
PubMed Links
Gene Details
- Gene
- Unc13b
- Gene Name
- unc-13 homolog B (C. elegans)
View Full Product Details