Product Details
- SNP ID
-
rs3706759
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.1:176782254 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCAGATCATGCAGACCACTTACCCA[C/T]GGGTTTCTCTTCAGATTTCAGTGCT
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Cep170
PubMed Links
Gene Details
- Gene
- Cep170
- Gene Name
- centrosomal protein 170
- Gene
- Mir350
- Gene Name
- microRNA 350
There are no transcripts associated with this gene.
View Full Product Details