Product Details

SNP ID
rs6262884
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.10:103169874 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
GGAAGCACAAAAACAGTTTCACATT[A/C]ATTCAAAATTAAATAATCCTAGGTA
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
Lrriq1 PubMed Links

Gene Details

Gene
Lrriq1
Gene Name
leucine-rich repeats and IQ motif containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001163559.1 Intron NP_001157031.1
NM_029134.2 Intron NP_083410.1

View Full Product Details