Product Details

SNP ID
rs13481116
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.13:102969331 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
ACCACATTTACTATCAAGTACTGGA[A/G]GATGCGCTGAACGATGACAGAAACC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
Mast4 PubMed Links

Gene Details

Gene
Mast4
Gene Name
microtubule associated serine/threonine kinase family member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_175171.3 Intron NP_780380.2

View Full Product Details