Product Details

SNP ID
rs13481650
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.12:111188138 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
ACTTGGACTCCTCAGTTCTCTTGAG[C/T]ATCCTTCAGATTCTTTACTCTGTAA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
4930595D18Rik PubMed Links

Gene Details

Gene
4930595D18Rik
Gene Name
RIKEN cDNA 4930595D18 gene
There are no transcripts associated with this gene.

Gene
Traf3
Gene Name
TNF receptor-associated factor 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286122.1 Intron NP_001273051.1
NM_011632.3 Intron NP_035762.2

View Full Product Details