Product Details
- SNP ID
-
rs13480828
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:128845964 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTTTGTCTCCTGCCCCTGCGGTCCC[C/T]CTCTTTTGTTTTCTCTTAAGCCTTT
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Sarnp
PubMed Links
Gene Details
- Gene
- Sarnp
- Gene Name
- SAP domain containing ribonucleoprotein
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_025364.2 |
|
Intron |
|
|
NP_079640.1 |
View Full Product Details