Product Details
- SNP ID
-
rs4166461
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:28404556 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAAAAATGCATTTTCTATATAAATA[A/G]CATCTTCTTCTTTCCAAAGAGCAGA
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Fgf12
PubMed Links
Gene Details
- Gene
- Fgf12
- Gene Name
- fibroblast growth factor 12
View Full Product Details