Product Details

SNP ID
rs13476296
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.1:189245633 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
AAAATGAAATACTGGCAGCTGGATG[A/G]TCTAAGAAGAAAAACACCAAGAAAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
Kcnk2 PubMed Links

Gene Details

Gene
Kcnk2
Gene Name
potassium channel, subfamily K, member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001159850.1 Intron NP_001153322.1
NM_001281847.1 Intron NP_001268776.1
NM_001281848.1 Intron NP_001268777.1
NM_010607.3 Intron NP_034737.2

View Full Product Details