Product Details
- SNP ID
-
rs6319386
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:18451096 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAAAAAAAAAAAAGCATAAAAATTT[C/T]AGTTTCCTCAGACAGCCTTTCTGTG
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Cttnbp2
PubMed Links
Gene Details
- Gene
- Cttnbp2
- Gene Name
- cortactin binding protein 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_080285.1 |
|
Intron |
|
|
NP_525024.1 |
View Full Product Details