Product Details

SNP ID
rs13477824
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.4:87224938 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
ACTTATAAATAATCAGAAAAGTGAG[A/G]TGACAGTCCTCTAATTAATCAGGTA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
Slc24a2 PubMed Links

Gene Details

Gene
Slc24a2
Gene Name
solute carrier family 24 (sodium/potassium/calcium exchanger), member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001110240.1 Intron NP_001103710.1
NM_172426.2 Intron NP_766014.1

View Full Product Details