Product Details
- SNP ID
-
rs6314295
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.13:20121190 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCTGGGGTTTACTGACTTGTTCAGA[A/G]TCTTCTCCTCTCTCCATATGACTGT
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Elmo1
PubMed Links
Gene Details
- Gene
- Elmo1
- Gene Name
- engulfment and cell motility 1
View Full Product Details