Product Details
- SNP ID
-
rs13476531
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:60441119 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TAAAATACAGCCACCCCTTGGGGCA[C/T]GTCCCAGGCTGTGTCGGCGGCTTCT
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Pla2r1
PubMed Links
Gene Details
- Gene
- Pla2r1
- Gene Name
- phospholipase A2 receptor 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_008867.2 |
3046 |
Silent Mutation |
|
|
NP_032893.1 |
View Full Product Details