Product Details
- SNP ID
-
rs13475897
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.1:65379118 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGAAACTTAGGTCCATCTGTGAACC[C/T]AGCCTTGGCTGCACCCTTGAGTTCA
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Pth2r
PubMed Links
Gene Details
- Gene
- Pth2r
- Gene Name
- parathyroid hormone 2 receptor
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_139270.2 |
|
Intron |
|
|
NP_644676.1 |
View Full Product Details