Product Details

SNP ID
rs4136858
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.10:84651735 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
GGTACCAGCCCTGTCTCCTCACCAT[A/G]GGTTTTTGAGAGGACGGTTCCTTGT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
Polr3b PubMed Links

Gene Details

Gene
Polr3b
Gene Name
polymerase (RNA) III (DNA directed) polypeptide B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_027423.1 Intron NP_081699.1

View Full Product Details