Product Details
- SNP ID
-
rs13483095
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:71897900 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGTAGTTGGGGTTGTAGTCGGTCAT[G/T]ATGGTCGAGGTCCGTAGCTTGCTCA
- Phenotype
-
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
Alk
PubMed Links
Gene Details
- Gene
- Alk
- Gene Name
- anaplastic lymphoma kinase
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_007439.2 |
3874 |
Silent Mutation |
|
|
NP_031465.2 |
View Full Product Details