Product Details
- SNP ID
-
rs4222751
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.1:162598718 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AATTCTGGTTTTACGATGTTGCATT[C/T]GATTCTTTTCACTTGAAAAGTCATG
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Vamp4
PubMed Links
Gene Details
- Gene
- Vamp4
- Gene Name
- vesicle-associated membrane protein 4
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_016796.3 |
2172 |
UTR 3 |
|
|
NP_058076.2 |
View Full Product Details