Product Details
- SNP ID
-
rs33049139
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:22524238 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACCTGTCAGAATACAGTGAATAAGT[A/G]TCTATGGAATGCTCCACCATAAATA
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Slc20a2
PubMed Links
Gene Details
- Gene
- Slc20a2
- Gene Name
- solute carrier family 20, member 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_011394.3 |
|
Intron |
|
|
NP_035524.2 |
View Full Product Details