Product Details
- SNP ID
-
rs4137502
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.1:30830773 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTTATTGAACTCTGTGCTCTGTAAC[C/T]GAGCATCATTAGACTCCGTGTCCTC
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Phf3
PubMed Links
Gene Details
- Gene
- Phf3
- Gene Name
- PHD finger protein 3
View Full Product Details