Product Details

SNP ID
rs13480933
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.11:29218539 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
AATCATCCCCTAGGAAAAGACCTCG[C/G]CTGGGCTCCTAGAATCCTTGCTGGG
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
Cfap36 PubMed Links

Gene Details

Gene
Cfap36
Gene Name
cilia and flagella associated protein 36
There are no transcripts associated with this gene.

Gene
Smek2
Gene Name
SMEK homolog 2, suppressor of mek1 (Dictyostelium)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_134034.2 2850 Silent Mutation NP_598795.2

View Full Product Details