Product Details
- SNP ID
-
rs4165480
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:25976392 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCAGACTCCCAGAGCTTGAATGGAT[G/T]CTGGCTACTTAGTTGTGCTGAACTG
- Phenotype
-
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
P3h2
PubMed Links
Gene Details
- Gene
- P3h2
- Gene Name
- prolyl 3-hydroxylase 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_173379.2 |
|
Intron |
|
|
NP_775555.1 |
View Full Product Details