Product Details
- SNP ID
-
rs6317022
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.1:176117187 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCTCCTTCTGACCTCTAACCACTTT[C/T]TAGGCCACAGTGCTGTTGCTGCCTC
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Pld5
PubMed Links
Gene Details
- Gene
- Pld5
- Gene Name
- phospholipase D family, member 5
View Full Product Details