Product Details
- SNP ID
-
rs13481566
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:84531872 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GACTGCGGGTTGGACGGGGCCAGGT[A/G]GGCCAAAGTAGTTGGCTCCTGGGAG
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Lin52
PubMed Links
Gene Details
- Gene
- Lin52
- Gene Name
- lin-52 homolog (C. elegans)
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_173756.4 |
|
Intron |
|
|
NP_776117.1 |
View Full Product Details