Product Details

SNP ID
rs4229986
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.13:95513896 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
TGAAGAGGATGGAGCAATACATGTT[A/G]CCATAGAAAAAGCCAATGAGCACCT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
F2rl1 PubMed Links

Gene Details

Gene
F2rl1
Gene Name
coagulation factor II (thrombin) receptor-like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007974.4 590 Silent Mutation NP_032000.3

View Full Product Details