Product Details

SNP ID
rs13483316
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.18:40324563 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
TATCTCTCTTTGGCATCAAGGCTGT[C/T]GTTTGAGCTGAATGGACAGTCATAA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
Kctd16 PubMed Links

Gene Details

Gene
Kctd16
Gene Name
potassium channel tetramerisation domain containing 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_026135.1 Intron NP_080411.1

View Full Product Details