Product Details
- SNP ID
-
rs4230834
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:76500735 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTAGTCCAGGGGACCCTAGGTCAAG[C/T]CACCACAACCCCAGTGGAGCACAGA
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Dgat1
PubMed Links
Gene Details
- Gene
- Dgat1
- Gene Name
- diacylglycerol O-acyltransferase 1
There are no transcripts associated with this gene.
- Gene
- Hsf1
- Gene Name
- heat shock factor 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_008296.2 |
1781 |
UTR 3 |
|
|
NP_032322.1 |
View Full Product Details