Product Details

SNP ID
rs13480819
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.10:127003427 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
CTTCAGGGCTGTGAGCAACGACCCC[C/G]GGATCATCACATGGAGAATTGAGGT
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
Avil PubMed Links

Gene Details

Gene
Avil
Gene Name
advillin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_009635.3 185 Missense Mutation NP_033765.2
Gene
Ctdsp2
Gene Name
CTD (carboxy-terminal domain, RNA polymerase II, polypeptide A) small phosphatase 2
There are no transcripts associated with this gene.

Gene
Mir26a-2
Gene Name
microRNA 26a-2
There are no transcripts associated with this gene.

Gene
Mir546
Gene Name
microRNA 546
There are no transcripts associated with this gene.

View Full Product Details