Product Details

SNP ID
rs13481763
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.13:36345315 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
TTTGATCAAGGATTTGCATATGATA[A/G]TTCTCTTTGGCCCACCCAAATAATG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
Fars2 PubMed Links

Gene Details

Gene
Fars2
Gene Name
phenylalanine-tRNA synthetase 2 (mitochondrial)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039189.2 Intron NP_001034278.1
NM_024274.3 Intron NP_077236.1

View Full Product Details