Product Details

SNP ID
rs4164241
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.16:14459042 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
CCCACAGTGGGGATACCAGCCAGCA[A/G]CACAGCAGCATAGCCGAACTGCAGA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
Abcc1 PubMed Links

Gene Details

Gene
Abcc1
Gene Name
ATP-binding cassette, sub-family C (CFTR/MRP), member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_008576.3 2810 Silent Mutation NP_032602.1

View Full Product Details