Product Details

SNP ID
rs6302685
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.17:70022752 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
CTCTATTCAAGGATTTGCTAAGGAG[A/C]GTTTGCTAGGCCATTCTGAGGCTTG
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
Dlgap1 PubMed Links

Gene Details

Gene
Dlgap1
Gene Name
discs, large (Drosophila) homolog-associated protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001128180.1 Intron NP_001121652.1
NM_001128181.1 Intron NP_001121653.1
NM_027712.3 Intron NP_081988.3
NM_177639.6 Intron NP_808307.2

View Full Product Details