Product Details
- SNP ID
-
rs13477633
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:33007129 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGGTCAGTAAGATTTATATTGCAAC[C/T]GACAGCACGCCGGTGGACATGCAGA
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Rragd
PubMed Links
Gene Details
- Gene
- Rragd
- Gene Name
- Ras-related GTP binding D
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_027491.2 |
1281 |
Silent Mutation |
|
|
NP_081767.2 |
View Full Product Details