Product Details

SNP ID
rs13481252
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.11:117246863 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
AGCTATGGGGATGTATGTGTATATA[C/T]CTCTCATTGTAAGTTCTGTATCTCC
Phenotype
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
Sept9 PubMed Links

Gene Details

Gene
Sept9
Gene Name
septin 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001113486.1 Intron NP_001106958.1
NM_001113487.1 Intron NP_001106959.1
NM_001113488.1 Intron NP_001106960.1
NM_017380.2 Intron NP_059076.1

View Full Product Details