Product Details

SNP ID
rs29819000
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.13:59704246 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
TGTACTCCAGGAGGAGGGTAGAAAC[A/G]AAGGCTCACAAGGTGGGCCTTTCAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
Spata31d1a PubMed Links

Gene Details

Gene
Spata31d1a
Gene Name
spermatogenesis associated 31 subfamily D, member 1A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_028157.2 Intron NP_082433.2
Gene
Spata31d1b
Gene Name
spermatogenesis associated 31 subfamily D, member 1B
There are no transcripts associated with this gene.

View Full Product Details