Product Details
- SNP ID
-
rs13477190
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:73049532 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCATAAGGTGATGCACTTGTTGGTC[C/T]CCCGTTAGGGTGGTAATCTCCAGGC
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Slitrk3
PubMed Links
Gene Details
- Gene
- Slitrk3
- Gene Name
- SLIT and NTRK-like family, member 3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_198864.2 |
2325 |
Missense Mutation |
|
|
NP_942564.2 |
View Full Product Details