Product Details

SNP ID
rs13481712
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.13:18138677 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
CTCTTCACTGACACAGGCATTCTCA[C/T]GTTGTGGCACAGGTGCGTATGCAGC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
Pou6f2 PubMed Links

Gene Details

Gene
Pou6f2
Gene Name
POU domain, class 6, transcription factor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_175006.2 Intron NP_778171.2

View Full Product Details