Product Details

SNP ID
rs6254148
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.X:75383429 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
AATAGTATATGTAAATGTATTTCTA[G/T]ACTTCTTATTCAACTAATTTTGTGT
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
F8 PubMed Links

Gene Details

Gene
F8
Gene Name
coagulation factor VIII
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001161373.1 Intron NP_001154845.1
NM_001161374.1 Intron NP_001154846.1
NM_007977.2 Intron NP_032003.2
Gene
Fundc2
Gene Name
FUN14 domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_026126.4 Intron NP_080402.3

View Full Product Details