Product Details

SNP ID
rs13481801
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.13:47083843 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
ATAAGCCATATGTTAAAAGTCTGAA[A/C]GTGGCAAAACTTACTCTTCCACTGA
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
Dek PubMed Links

Gene Details

Gene
Dek
Gene Name
DEK oncogene (DNA binding)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_025900.2 3551 Intron NP_080176.2
Gene
Kdm1b
Gene Name
lysine (K)-specific demethylase 1B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_172262.3 3551 UTR 3 NP_758466.1

View Full Product Details