Product Details
- SNP ID
-
rs6313863
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.13:47072684 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CACAGAGCCCTGTTCTTGTAACCCA[A/G]GCTGACCCACTTGCTACATGGCTCT
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Kdm1b
PubMed Links
Gene Details
- Gene
- Kdm1b
- Gene Name
- lysine (K)-specific demethylase 1B
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_172262.3 |
|
Intron |
|
|
NP_758466.1 |
View Full Product Details