Product Details

SNP ID
rs13481263
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.11:119839122 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
CACCAGGCACAGCATGGTGACTCTG[C/T]TCTTGAGGGTCATCTTTCACACAGG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
Rptor PubMed Links

Gene Details

Gene
Rptor
Gene Name
regulatory associated protein of MTOR, complex 1
There are no transcripts associated with this gene.

Gene
Rptoros
Gene Name
regulatory associated protein of MTOR, complex 1, opposite strand
There are no transcripts associated with this gene.

View Full Product Details