Product Details

SNP ID
rs13482997
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.17:43824637 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
TACTCGAAACACAATTTCTTGATGA[C/G]GGTATTCAGGTAGGCTTGTGGAAAT
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
Rcan2 PubMed Links

Gene Details

Gene
Rcan2
Gene Name
regulator of calcineurin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286653.1 Intron NP_001273582.1
NM_001286654.1 Intron NP_001273583.1
NM_030598.2 Intron NP_085101.1
NM_207649.1 Intron NP_997532.1

View Full Product Details