Product Details

SNP ID
rs13482437
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.15:12834339 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
CCCCTGGGTACTCGCACAGCTTCCC[A/G]CCGCCCGGCTTCAACAGTTACCAGA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
6030458C11Rik PubMed Links

Gene Details

Gene
6030458C11Rik
Gene Name
RIKEN cDNA 6030458C11 gene
There are no transcripts associated with this gene.

Gene
Drosha
Gene Name
drosha, ribonuclease type III
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130149.1 804 Silent Mutation NP_001123621.1
NM_026799.3 804 Silent Mutation NP_081075.3

View Full Product Details