Product Details

SNP ID
rs13481010
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.11:46817658 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
CAACAGTCATGACCACATCTGTTCT[C/T]CCAACCACCACACCACCCCAGACAC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
Timd4 PubMed Links

Gene Details

Gene
Timd4
Gene Name
T cell immunoglobulin and mucin domain containing 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178759.4 523 Silent Mutation NP_848874.3

View Full Product Details